Skip to Content
Merck
All Photos(1)

Key Documents

A1880

Sigma-Aldrich

(±)-2-Aminopimelic acid

≥99%

Synonym(s):

(±)-2-Aminoheptanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO4
CAS Number:
Molecular Weight:
175.18
Beilstein:
1722015
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99%

SMILES string

NC(CCCCC(O)=O)C(O)=O

InChI

1S/C7H13NO4/c8-5(7(11)12)3-1-2-4-6(9)10/h5H,1-4,8H2,(H,9,10)(H,11,12)

InChI key

JUQLUIFNNFIIKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

2-Aminopimelic acid may be used to create peptides that are antibacterial agents that inhibit diaminopimelic acid biosynthesis required for cell wall biosynthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Berges et al.
Journal of medicinal chemistry, 29(1), 89-95 (1986-01-01)
Succinyl-CoA:tetrahydrodipicolinate-N-succinyltransferase is a key enzyme in the biosynthesis of diaminopimelic acid (DAP), a component of the cell wall peptidoglycan of nearly all bacteria. This enzyme converts the cyclic precursor tetrahydrodipicolinic acid (THDPA) to a succinylated acyclic product. L-2-Aminopimelic acid (L-1)
P Le Roux et al.
International journal of peptide and protein research, 37(2), 103-111 (1991-02-01)
Taking advantage of the peptide transport strategy, we have designed and synthesized several new peptides containing 2-aminopimelic acid (Apm), an inhibitor of the diaminopimelate pathway in bacteria: L-Lys-ambo-Apm, ambo-Apm-L-Lys, L-Lys-L-Ala-ambo-Apm, ambo-Apm-L-Ala-L-Lys, L-Ala(Cl)-ambo-Apm and ambo-Apm-L-Ala(Cl). In the two latter cases, Apm
Gabriele Netzel et al.
Toxins, 11(3) (2019-03-22)
In vitro experiments have demonstrated that camel foregut-fluid has the capacity to metabolize indospicine, a natural toxin which causes hepatotoxicosis, but such metabolism is in competition with absorption and outflow of indospicine from the different segments of the digestive system.
Jie Zhao et al.
Biomedical chromatography : BMC, 32(10), e4299-e4299 (2018-05-26)
Triptolide, a major active constitute of Tripterygium wilfordii Hook. F, is prescribed for the treatment of autoimmune diseases in China. One of its most severe adverse effects observed in the clinical use is hepatotoxicity, but the mechanism is still unknown.
Hui-Chang Bi et al.
Carcinogenesis, 35(10), 2264-2272 (2014-08-15)
The cell metabolome comprises abundant information that may be predictive of cell functions in response to epigenetic or genetic changes at different stages of cell proliferation and metastasis. An unbiased ultra-performance liquid chromatography-mass spectrometry-based metabolomics study revealed a significantly altered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service