Skip to Content
Merck
All Photos(1)

Key Documents

89870

Sigma-Aldrich

Toluene 2,4-diisocyanate

purum, suitable for electron microscopy, ≥98.0% (GC)

Synonym(s):

2,4-Diisocyanatotoluene, 4-Methyl-m-phenylene diisocyanate, Tolylene 2,4-diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2O2
CAS Number:
Molecular Weight:
174.16
Beilstein:
744602
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25
form:
liquid
Assay:
≥98.0% (GC)
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

liquid

technique(s)

electron microscopy: suitable

refractive index

n20/D 1.567

bp

124-126 °C/18 mmHg (lit.)
251 °C (lit.)

mp

19.5-21.5 °C (lit.)

density

1.21 g/mL at 20 °C
1.22 g/mL at 20 °C (lit.)

SMILES string

Cc1ccc(cc1N=C=O)N=C=O

InChI

1S/C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3

InChI key

DVKJHBMWWAPEIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Toluene 2,4-diisocyanate is used to study it occupational hazards (asthma, dermatitis) and mechanisms of action. Reagent for covalent protein crosslinking[1]; For the immobilization of antibodies to a plastic surface for RIA[2]

Other Notes

Reagent for covalent protein crosslinking[1]; For the immobilization of antibodies to a plastic surface for RIA[2]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Saito et al.
Clinica chimica acta; international journal of clinical chemistry, 133(3), 301-310 (1983-10-14)
A novel procedure to fix antibody on a plastic surface was developed, using a polystyrene tube treated with 0.5% toluene 2,4-diisocyanate (TC) dissolved in carbon tetrachloride. Using 125I-labelled IgG, we found that the rate of immobilization of IgG was significantly
Shuyu Chen et al.
Allergy, asthma & immunology research, 12(4), 608-625 (2020-05-14)
Accumulating evidence has suggested that toll-like receptor 4 (TLR4) is critically involved in the pathogenesis of asthma. The aim of this study was to investigate the role of TLR4 in toluene diisocyanate (TDI)-induced allergic airway inflammation. TLR4-/- and wild-type (WT)
[57] Bifunctional reagents.
F Wold
Methods in enzymology, 25, 623-651 (1972-01-01)
Rahul Purwar et al.
Experimental dermatology, 20(9), 709-714 (2011-05-17)
Keratinocytes synthesize complement component 3 (C3) constitutively, and increased expression of C3 has been described during skin inflammation. In this study, we investigated the role of C3 in T cell-mediated allergic contact dermatitis, which is a clinical manifestation of contact
Olov Collste et al.
Journal of cardiac failure, 20(12), 968-972 (2014-10-01)
Our objective was to investigate whether dobutamine stress echocardiography (DSE) could induce abnormal cardiac function in takotsubo stress cardiomyopathy (TSC) patients in a stable condition after the acute attack. This was a case-control study and a substudy of the Stockholm

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service