Skip to Content
Merck
All Photos(1)

Key Documents

12097

Sigma-Aldrich

Benzethonium hydroxide solution

suitable for tissue culture, ~1.0 M in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO3
CAS Number:
Molecular Weight:
429.64
Beilstein:
3915309
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

concentration

~1.0 M in methanol

technique(s)

tissue culture: suitable

storage temp.

2-8°C

SMILES string

[OH-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

InChI

1S/C27H42NO2.H2O/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H2/q+1;/p-1

InChI key

USLZJBWCHRRIAQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Benzethonium hydroxide has been used in a study to evaluate the kinetics of (14)carbon dioxide ((14)CO(2)) excretion by oral commensal flora to theoretically propose optimum breath collection timings for (14) carbon urea breath test.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chander M Pathak et al.
Journal of gastroenterology and hepatology, 23(10), 1603-1607 (2008-05-01)
Previous studies have shown that while performing the (14)C-urea breath test ((14)C-UBT) for the detection of Helicobacter pylori (H. pylori), there is possibility of false-positive results due to the other urease producing bacteria present in oropharynx, if breath samples are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service