Skip to Content
Merck
All Photos(1)

Documents

Y0001626

Ziprasidone mesylate trihydrate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25ClN4O4S2 · 3H2O
CAS Number:
Molecular Weight:
563.09
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ziprasidone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C21H21ClN4OS.CH4O3S.3H2O/c22-17-13-18-15(12-20(27)23-18)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)28-24-21;1-5(2,3)4;;;/h1-4,11,13H,5-10,12H2,(H,23,27);1H3,(H,2,3,4);3*1H2

InChI key

WLQZEFFFIUHSJB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ziprasidone mesylate trihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziprasidone malate, a new trimorphic salt with improved aqueous solubility.
Portell A, et al.
CrystEngComm, 11(5), 791-795 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service