Skip to Content
Merck
All Photos(1)

Key Documents

Y0000717

Moxifloxacin for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Moxifloxacin hydrochloride, 1-Cyclopropyl-6-fluoro-8-methoxy-7-[(4aS,7aS)-octahydropyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24FN3O4· HCl
CAS Number:
Molecular Weight:
437.89
Beilstein:
8377447
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

moxifloxacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.COc1c(N2C[C@@H]3CCCN[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(O)=O

InChI

1S/C21H24FN3O4.ClH/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24;/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28);1H/t11-,16+;/m0./s1

InChI key

IDIIJJHBXUESQI-DFIJPDEKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Moxifloxacin for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohit Shetty et al.
BioMed research international, 2014, 340509-340509 (2014-10-11)
To report the profile of microbial keratitis occurring after corneal collagen cross-linking (CXL) in keratoconus patients. A retrospective analysis of 2350 patients (1715 conventional CXL, 310 transepithelial CXL, and 325 accelerated CXL) over 7 years (from January 2007 to January
Cemile Banu Cosar et al.
European journal of ophthalmology, 24(3), 325-329 (2013-09-24)
To investigate refractive and visual outcomes and complications of the Supracor procedure to treat presbyopia. A total of 123 eyes from 68 presbyopic patients with hyperopia were enrolled in this prospective, nonrandomized, clinical trial. The Supracor procedure was performed using
Lingmin He et al.
JAMA ophthalmology, 133(1), 51-59 (2014-10-17)
Wavefront-guided (WFG) and wavefront-optimized (WFO) platforms for refractive surgery are designed for improved visual outcomes. It is unclear which treatment profile is superior for patients undergoing photorefractive keratectomy (PRK). To compare the safety, efficacy, predictability, stability, and higher-order aberrations in
Sumeet Khanduja et al.
Journal of glaucoma, 24(4), 267-271 (2013-01-23)
To evaluate the change in anterior chamber angle parameters after scleral buckling surgery using anterior segment optical coherence tomography (ASOCT). Fifty-five phakic eyes of 55 subjects undergoing scleral buckling with the placement of an encircling band for primary rhegmatogenous retinal
Lingmin He et al.
American journal of ophthalmology, 157(6), 1170-1178 (2014-02-25)
To compare the clinical outcomes of wavefront-guided and wavefront-optimized laser in situ keratomileusis (LASIK). Prospective, randomized, fellow-eye-controlled study. The setting was a single academic institution. The study population included 110 eyes of 55 patients with myopia with and without astigmatism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service