Skip to Content
Merck
All Photos(2)

Key Documents

W429420

Sigma-Aldrich

Ethyl dimethyl dioxolane acetate

mixture of isomers, ≥97%, FG

Synonym(s):

fruity ketal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O4
CAS Number:
Molecular Weight:
188.22
FEMA Number:
4294
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.087
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.428 (lit.)

density

1.042 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; fruity

SMILES string

CCOC(=O)CC1(C)OCC(C)O1

InChI

1S/C9H16O4/c1-4-11-8(10)5-9(3)12-6-7(2)13-9/h7H,4-6H2,1-3H3

InChI key

GSIXJEIRJVOUFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl dimethyl dioxolane acetate(ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate) is an isomeric mixture of ethyl dimethyl dioxolane acetate. It is also referred to as ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service