Skip to Content
Merck
All Photos(1)

Documents

SMB00929

Sigma-Aldrich

5′-Hydroxy carvedilol

≥95% (HPLC)

Synonym(s):

5′-Hydroxy carvedilol, 3-[2-[[3-(9H-Carbazol-4-yloxy)-2-hydroxypropyl]amino]ethoxy]-4-methoxyphenol, 5′-Hydroxyphenyl carvedilol, 5′-Hydroxyphenylcarvedilol, 5′-OH carvedilol, 5-Hydroxycarvedilol, 5OHC, BM-140830, Carvedilol metabolite M5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O5
CAS Number:
Molecular Weight:
422.47
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥95% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble, clear, colorless

storage temp.

2-8°C

InChI

1S/C24H26N2O5/c1-29-21-10-9-16(27)13-23(21)30-12-11-25-14-17(28)15-31-22-8-4-7-20-24(22)18-5-2-3-6-19(18)26-20/h2-10,13,17,25-28H,11-12,14-15H2,1H3

InChI key

PVUVZUBTCLBJMT-UHFFFAOYSA-N

General description

Carvedilol is a β- and α1-adrenoreceptor blocker for the treatment of hypertension and congestive heart failure (CHF). The drug is metabolized by CYP2D6 (to 4′-OH and 5′-OH), CYP2C9 (to O-desmethyl), CYP1A2 (to 8-OH). It is also oxidized to 1-OH carvedilol, but the enzyme involved is not yet clear. These metabolites are useful markers for studying and monitoring the activities of cytochrome metabolizing enzymes. M4 metabolite (4′-OH), but not M2 or M5, is most likely to contribute to total β-adrenoceptor blocking activity due to its higher potency.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Target Organs

Liver,spleen,Adrenal gland,Uterus (including cervix)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W H Schaefer et al.
Drug metabolism and disposition: the biological fate of chemicals, 26(10), 958-969 (1998-10-08)
The excretion and biotransformation of carvedilol [1-[carbazolyl-(4)-oxy]-3-[(2-methoxyphenoxyethyl)amino]-2-p ropanol], a new, multiple-action, neurohormonal antagonist that exhibits the combined pharmacological activities of beta-adrenoreceptor antagonism, vasodilation, and antioxidation, were investigated in dogs, rats, and mice. Carvedilol was absorbed well, and biliary secretion was
D Tenero et al.
Journal of clinical pharmacology, 40(8), 844-853 (2000-08-10)
Carvedilol is a relatively new drug with beta- and alpha 1-receptor blocking activity and antioxidant effects recently approved for the treatment of congestive heart failure (CHF). An ascending, multiple-dose study was completed in 20 male patients with stable New York
Eben Jung et al.
Journal of Korean medical science, 33(27), e182-e182 (2018-07-03)
Carvedilol is commonly used to treat hypertension as a β- and α1-adrenoreceptor blocker, but it is metabolized by CYP2D6, and CYP2D6*10 allele is dominant in Asian population. The objective of this study was to assess the influence of CYP2D6 polymorphisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service