Skip to Content
Merck
All Photos(1)

Documents

PFS15

Supelco

SpheriCal Mix

Protein Medium, suitable for MALDI-MS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

product line

SpheriCal

form

powder

shelf life

expiry date on the label

mol wt

Mr 15000-30000 Da (Na & adduct)

composition

9-nitroanthracene, >90%
sodium trifluoroacetate, <10%

technique(s)

MALDI-MS: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

calibrants PFS5A : PFS5B : PFS5C : PFS5D = 1:1:1:1
Matrix/counter ion: 9-Nitroanthracene/Na+

Application

For mass calibration in mass spectrometry

Packaging

Quantity: 3*5 mg

Legal Information

SpheriCal is a trademark of Polymer Factory Sweden AB

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MALDI-TOF MS is a tool for confirming end group identity & transformations, verifying successful & complete conjugation. Functionalized polymers enable a quick & efficient route for synthesis of various polymer conjugates for many applications if apt end group is present.

Related Content

The Grayson research labs have focused on the synthesis and characterization of well-defined polymers. One particular theme of research has involved the optimization of the mass spectral analysis of polymers using MALDI-TOF MS and associated calibration methodologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service