Skip to Content
Merck
All Photos(1)

Documents

NIST8650

Ground Kudzu (Pueraria Montan Var. Lobata) Rhizome

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 15 g

manufacturer/tradename

NIST®

application(s)

agriculture

format

matrix material

General description

One unit of RM 8650 contains five packets, each comprising about 3 grams of kudzu rhizome powder which can be stored at room temperature.
For more information, please refer to the SDS and COA.
SRM 1993_cert
SRM 1993_SDS

Application
Ground Kudzu Rhizome Reference Material is intended for evaluating analytical methods used to determine isoflavones and components in kudzu rhizomes and similar matrices.

Features and Benefits

  • The reference mass fraction values forisoflavones and elements in RM 8650 are provided on a dry-mass basis and valuesare determinedby inductivelycoupled plasma mass spectrometry (ICP-MS).
  • NIST certificate is providedwith expirationdetails and instructions for storage and use.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service