Skip to Content
Merck
All Photos(1)

Documents

NIST2619A

Carbon dioxide in nitrogen

NIST® SRM® 2619a, nominal 0.5% mol/mol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352119

grade

certified reference material

Quality Level

packaging

pkg of cylinder

manufacturer/tradename

NIST®

application(s)

air monitoring
environmental

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a primary gas mixture for which the amount-of-substance fraction, expressed as concentration, may be related to secondary working standards. This SRM is intended for the calibration of instruments used for carbon dioxide determinations and for other applications.

This SRM mixture is supplied in a DOT 3AL-specification aluminum (6061 alloy) cylinder with a water volume of 6 L. Mixtures are shipped with a nominal pressure exceeding 12.4 MPa (1800 psig), which provides the user with 0.73 m3 (25.8 ft3) of useable mixture. The cylinder is the property of the purchaser and is equipped with a CGA-580 brass valve, which is the recommended outlet for this carbon dioxide mixture. For more information, please refer to the SDS and COA.

SRM 2619A_cert

SRM 2619A_SDS

Other Notes

Certified for the analytes listed below.
Carbon Dioxide (CO2)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service