Skip to Content
Merck
All Photos(1)

Key Documents

D83303

Sigma-Aldrich

Diethanolamine

99%

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

Assay

99%

form

solid

autoignition temp.

689 °F

expl. lim.

10.6 %

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esmeralda García-Díaz et al.
Water research, 183, 116095-116095 (2020-07-10)
Photocatalytic water treatment has significant potential to disinfect and degrade recalcitrant organic pollutants while minimizing the need to add chemicals, but current approaches have poor energy efficiency due, in part, to inefficient utilization of photo-generated reactive oxygen species (ROS). Organic
Danning Zhang et al.
Environmental science & technology, 52(21), 12402-12411 (2018-10-03)
Micrometer-sized titanium dioxide hierarchical spheres (TiO2-HS) were assembled from nanosheets to address two common limitations of photocatalytic water treatment: (1) inefficiency associated with scavenging of oxidation capacity by nontarget water constituents and (2) energy-intensive separation and recovery of the photocatalyst
Bjørn Henrik Hansen et al.
Aquatic toxicology (Amsterdam, Netherlands), 99(2), 212-222 (2010-06-12)
Alkanolamines are surface-active chemicals used in a wide range of industrial, agricultural and pharmaceutical applications and products. Of particular interest is the use of alkanolamines such as diethanolamine (DEA) in the removal of CO(2) from natural gas and for CO(2)
Wenting Zhang et al.
PloS one, 10(5), e0125995-e0125995 (2015-05-15)
We previously reported a new approach for culturing difficult-to-preserve primary patient-derived multiple myeloma cells (MMC) using an osteoblast (OSB)-derived 3D tissue scaffold constructed in a perfused microfluidic environment and a culture medium supplemented with patient plasma. In the current study
Shiguo Sun et al.
Dalton transactions (Cambridge, England : 2003), (38)(38), 7969-7974 (2009-09-23)
A series of ruthenium(II) tris-bipyridyl complexes covalently linked with different amine reductants such as tripropylamine (TPrA), ethanolamine and diethanolamine for an electrochemiluminescence (ECL) system have been synthesized. Their ECL property at different working electrodes has been studied with and without

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service