D15006
1,8-Diamino-4,5-dihydroxyanthraquinone
technical grade, 90%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H10N2O4
CAS Number:
Molecular Weight:
270.24
EC Number:
MDL number:
UNSPSC Code:
12171500
grade
technical grade
Assay
90%
mp
>300 °C (lit.)
SMILES string
Nc1ccc(O)c2C(=O)c3c(O)ccc(N)c3C(=O)c12
InChI
1S/C14H10N2O4/c15-5-1-3-7(17)11-9(5)13(19)10-6(16)2-4-8(18)12(10)14(11)20/h1-4,17-18H,15-16H2
InChI key
SJHHHHHQWQOCDQ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analytica Chimica Acta, 74, 441-441 (1975)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service