Skip to Content
Merck
All Photos(1)

Documents

BCR288B

Lead with added impurities (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

General description

Certified for the mass fractions of As, Bi, Cd, Cu, Se, Te, Tl and Zn.

Analysis Note

For more information please see:
BCR288B

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service