Skip to Content
Merck
All Photos(1)

Documents

93383

Supelco

2,4,6-Tris(pentafluoroethyl)-1,3,5-triazine

analytical standard, suitable for mass spectrometry (MS)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9F15N3
CAS Number:
Molecular Weight:
435.09
Beilstein:
371291
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Assay

≥98.0% (NMR)

technique(s)

mass spectrometry (MS): suitable

refractive index

n20/D 1.312

density

1.665 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)c1nc(nc(n1)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chemical mass marker for negative ion chemical ionization mass spectrometry

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.V. Bowen et al.
Analytical Chemistry, 47, 2289-2289 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service