Skip to Content
Merck
All Photos(1)

Documents

90609

Supelco

Vanillylidenacetone

analytical standard

Synonym(s):

4-(4-Hydroxy-3-methoxyphenyl)-3-buten-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

SMILES string

COc1cc(\C=C\C(C)=O)ccc1O

InChI

1S/C11H12O3/c1-8(12)3-4-9-5-6-10(13)11(7-9)14-2/h3-7,13H,1-2H3/b4-3+

InChI key

AFWKBSMFXWNGRE-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Bock et al.
Journal of Raman spectroscopy : JRS, 50(6), 778-792 (2019-07-03)
Anatomical and chemical information can be linked by Raman imaging. Behind every pixel of the image is a Raman spectrum, which contains all the information as a molecular fingerprint. Yet to understand the spectra, the bands have to be assigned

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service