Skip to Content
Merck
All Photos(1)

Key Documents

88295

Sigma-Aldrich

Thallium(III) trifluoroacetate

technical, ~42% Tl basis (RT)

Synonym(s):

Thallic trifluoroacetate, Trifluoroacetic acid thallium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3COO)3Tl
CAS Number:
Molecular Weight:
543.43
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

grade

technical

concentration

~42% (Tl, RT)

impurities

Tl2O3

mp

213 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)O[Tl](OC(=O)C(F)(F)F)OC(=O)C(F)(F)F

InChI

1S/3C2HF3O2.Tl/c3*3-2(4,5)1(6)7;/h3*(H,6,7);/q;;;+3/p-3

InChI key

PSHNNUKOUQCMSG-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pamela R Hall et al.
Antimicrobial agents and chemotherapy, 52(6), 2079-2088 (2008-04-09)
Viral entry into susceptible host cells typically results from multivalent interactions between viral surface proteins and host entry receptors. In the case of Sin Nombre virus (SNV), a New World hantavirus that causes hantavirus cardiopulmonary syndrome, infection involves the interaction
Studies on peptides. CLVI. Synthesis of second human calcitonin gene-related peptide (beta-hCGRP) by application of a new disulfide-bonding reaction with thallium (III) trifluoroacetate.
N Fujii et al.
Chemical & pharmaceutical bulletin, 35(12), 4769-4776 (1987-12-01)
Studies on peptides. CLI. Syntheses of cystine-peptides by oxidation of S-protected cysteine-peptides with thallium(III) trifluoroacetate.
N Fujii et al.
Chemical & pharmaceutical bulletin, 35(6), 2339-2347 (1987-06-01)
Karen Page et al.
Journal of peptide science : an official publication of the European Peptide Society, 13(12), 833-838 (2007-08-30)
Human amylin (1-37) and the (1-13) fragment were synthesized with and without pseudoproline dipeptides. Thallium (III) trifluoroacetate, a mild oxidant, was used to cyclize the peptides by forming a disulfide bridge from C(2) to C(7). On the basis of our
Jianmin Chen et al.
Toxicology, 222(3), 195-201 (2006-04-06)
Particulate air pollution is associated with respiratory and cardiovascular morbidity and mortality. However, important uncertainties remain in the quantification of extrapulmonary translocation of ultrafine particles into blood circulation. Therefore, the widely used radioiodinated technique was applied to radiolabel polystyrene particles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service