Skip to Content
Merck
All Photos(1)

Key Documents

86934

Supelco

Tetrabutylphosphonium tetrafluoroborate

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4P(BF4)
CAS Number:
Molecular Weight:
346.24
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

≥99.0% (T)
≥99.0%

conductivity measuring range

~19.2 mS/cm (from 0.1 g/ml acetonitrile)

mp

96-99 °C

solubility

methanol: 0.1 g/mL, clear, colorless

SMILES string

F[B-](F)(F)F.CCCC[P+](CCCC)(CCCC)CCCC

InChI

1S/C16H36P.BF4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3,4)5/h5-16H2,1-4H3;/q+1;-1

InChI key

BYCHYJSDSHOWIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Novel organic electrolyte with high conductivity in high energy density electric double-layer capacitors (EDLC)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piyush Sindhu Sharma et al.
Biosensors & bioelectronics, 100, 251-258 (2017-09-22)
By means of molecular imprinting of a conducting polymer, molecular cavities selective for oxytocin nonapeptide, an autism biomarker, were designed. Embedding of the oxytocin template, and then its extracting from the molecularly imprinted polymer (MIP) was confirmed by the XPS
M. Ue et al.
Journal of the Electrochemical Society, 141, 2989-2989 null
Ya-Fei Lu et al.
Oncogene, 39(24), 4650-4665 (2020-05-15)
Sirtuin 7 (SIRT7), an NAD+-dependent deacetylase, plays vital roles in energy sensing, but the underlying mechanisms of action remain less clear. Here, we report that SIRT7 is required for p53-dependent cell-cycle arrest during glucose deprivation. We show that SIRT7 directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service