84838
Dibutyl sebacate
Selectophore™, ≥97.0%
Synonym(s):
Decanedioic acid dibutyl ester, Sebacic acid dibutyl ester
About This Item
Recommended Products
grade
for ion-selective electrodes
Quality Level
product line
Selectophore™
Assay
≥97.0% (GC)
≥97.0%
form
liquid
refractive index
n20/D 1.441 (lit.)
n20/D 1.441
bp
178-179 °C/3 mmHg (lit.)
solubility
THF: 1 g/10 mL
density
0.936 g/mL at 25 °C (lit.)
functional group
ester
SMILES string
CCCCOC(=O)CCCCCCCCC(=O)OCCCC
InChI
1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3
InChI key
PYGXAGIECVVIOZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
366.8 °F - open cup
Flash Point(C)
186 °C - open cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service