Skip to Content
Merck
All Photos(1)

Key Documents

80012

Supelco

2,3,5,6-Tetrabromo-p-xylene

analytical standard

Synonym(s):

1,2,4,5-Tetrabromo-3,6-dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Br4(CH3)2
CAS Number:
Molecular Weight:
421.75
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

mp

254-256 °C (lit.)

application(s)

environmental

SMILES string

Cc1c(Br)c(Br)c(C)c(Br)c1Br

InChI

1S/C8H6Br4/c1-3-5(9)7(11)4(2)8(12)6(3)10/h1-2H3

InChI key

RXKOKVQKECXYOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service