Skip to Content
Merck
All Photos(1)

Key Documents

79170

Sigma-Aldrich

3-Phenylserine monohydrate

puriss., ≥99.0% (NT)

Synonym(s):

β-Hydroxyphenylalanine, 2-Amino-3-hydroxy-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO3 · H2O
CAS Number:
Molecular Weight:
199.20
Beilstein:
2102175
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

puriss.

Assay

≥99.0% (NT)

mp

202-208 °C (dec.)

SMILES string

O.NC(C(O)c1ccccc1)C(O)=O

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service