Skip to Content
Merck
All Photos(1)

Key Documents

76646

Supelco

Saflufenacil Metabolite M800H35

analytical standard

Synonym(s):

N-[4-Chloro-2-fluoro-5-({[(isopropylamino)sulfonyl]amino}carbonyl)phenyl]urea, 5-[(Aminocarbonyl)amino]-2-chloro-4-fluoro-N-[[(1-methylethyl)amino]sulfonyl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14ClFN4O4S
CAS Number:
Molecular Weight:
352.77
Enzyme Commission number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(NS(NC(C)C)(=O)=O)C1=C(Cl)C=C(F)C(NC(N)=O)=C1

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service