Skip to Content
Merck
All Photos(1)

Key Documents

75110

Supelco

Phyllanthin

analytical standard

Synonym(s):

(2S,3S)-(+)-1,4-Dimethoxy-2,3-diveratrylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O6
CAS Number:
Molecular Weight:
418.52
Beilstein:
2066503
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COC[C@@H](Cc1ccc(OC)c(OC)c1)[C@@H](COC)Cc2ccc(OC)c(OC)c2

InChI

1S/C24H34O6/c1-25-15-19(11-17-7-9-21(27-3)23(13-17)29-5)20(16-26-2)12-18-8-10-22(28-4)24(14-18)30-6/h7-10,13-14,19-20H,11-12,15-16H2,1-6H3/t19-,20-/m1/s1

InChI key

KFLQGJQSLUYUBF-WOJBJXKFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phyllanthin is found in many Phyllanthus species. It is a lignan compound which exhibits various biochemical and pharmacological properties.[1][2]

Application

Phyllanthin may be used as a reference standard for the determination of the analyte in Phyllanthus species by high-performance thin layer chromatography (HPTLC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective activity of phyllanthin in ethanol-treated primary culture of rat hepatocytes.
Chirdchupunseree H and Pramyothin P
Journal of Ethnopharmacology, 128(1), 172- 176 (2010)
Phyllanthin from Phyllanthus amarus inhibits cellular and humoral immune responses in Balb/C mice.
Ilangkovan M, et al.
Phytomedicine, 23(12), 1441- 1450 (2016)
Quantitative determination of phyllanthin and hypophyllanthin in Phyllanthus species by high-performance thin layer chromatography.
Tripathi AK, et al.
Phytochemical Analysis, 17(6), 394- 397 (2006)
Cong Zhao et al.
Environmental toxicology (2020-08-17)
Ischemic heart disease is an imperative cause of high morbidity and mortality globally. The cardiac ischemia/reperfusion damage occur in both reperfusion and ischemia. In this exploration, we have planned to examine the cardio-protective action of phyllanthin against the myocardial ischemic-reperfusion
Nguyen Duc Hanh et al.
Drug development and industrial pharmacy, 40(6), 793-802 (2013-04-19)
Phyllanthin is a major bioactive lignan component of Phyllanthus amarus, with several known biological activities. This study dealt with the isolation and physicochemical characterization of phyllanthin. Phyllanthin was isolated from P. amarus leaves by column chromatography and purified by recrystallization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service