Skip to Content
Merck
All Photos(1)

Documents

73831

Millipore

Tinsdale Supplement

suitable for microbiology

Synonym(s):

Diphtheria Virulence Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Corynebacterium spp.

Components

(1 vial of each is sufficient for 1000 mL medium)
Tinsdale Supplement part A (Cat. No. 74933): horse serum 100ml
Tinsdale Supplement part B (Cat. No. 66784): potassium tellurite (1% after diluting in 30 ml water) 1ml

Analysis Note

Tinsdale Supplement is used together with Tinsdale Agar Base for the primary isolation and identification of Corynebacterium diptheriae.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service