Skip to Content
Merck
All Photos(1)

Documents

70442

Sigma-Aldrich

1-Naphthol

≥98.0% (GC)

Synonym(s):

α-Naphthol, 1-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein:
1817321
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.5 (120 °C, vs air)

vapor pressure

1 mmHg ( 94 °C)

Assay

≥98.0% (GC)

autoignition temp.

1007 °F

expl. lim.

5 %

bp

278-280 °C (lit.)

mp

94-96 °C (lit.)
94-96 °C

SMILES string

Oc1cccc2ccccc12

InChI

1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H

InChI key

KJCVRFUGPWSIIH-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

1-Naphthol is the hydroxyl derivative of naphthalene. It forms hydrogen bonded complexes with H2O, CH3OH, NH3 and ND3 in gas phase. Hydrogen-bond dissociation energy of these complexes has been evaluated. Its sorption by biochars (obtained from orange peels, having pyrolytic temperatures in the range of 150-700 °C and designated as OP150-OP700) has been studied. It has been identified as an impurity in the active pharmaceutical ingredient (API) of duloxetine hydrochloride.

Application

1-Naphthol (Naphthalen-1-ol) may be used as a standard for the NMR, FT-IR and mass spectrometric characterization of impurities present in the Duloxetine Hydrochloride active pharmaceutical ingredient (API).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 2 Oral - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accurate hydrogen-bonding energies between 1-naphthol and water, methanol and ammonia.
Burgi T, et al
Chemical Physics Letters, 246(3), 291-299 (1995)
Characterization of duloxetine HCL API and its process related impurities
Karagiannidou E, et al.
International Journal of Analytical, Pharmaceutical and Biomedical Sciences, 3(2) (2014)
Baoliang Chen et al.
Chemosphere, 76(1), 127-133 (2009-03-14)
Biochars, derived from biomass, are increasingly recognized as an environmental-friendly sorbent to abate organic pollutants. Sorption variations of biochars with their pyrolytic temperatures are evaluated. Nine biochars of orange peels with different pyrolytic temperatures (150-700 degrees C, referred as OP150-OP700)
Anika Kasprick et al.
European journal of immunology, 45(5), 1462-1470 (2015-02-14)
The role of mast cells (MCs) in autoimmunity is the matter of an intensive scientific debate. Based on observations in different MC-deficient mouse strains, MCs are considered as fundamental players in autoimmune diseases. However, most recent data suggest that the
Hongbo Zhang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(8), 1286-1293 (2012-01-24)
In vitro glucuronidation assays of diclofenac and indomethacin at pH 7.4 are biased by the instability of the glucuronides due to acyl migration. The extent of this acyl migration may be reduced significantly by performing the glucuronidation reaction at pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service