Key Documents
64013BC
Millipore
MVP ICON® Field Care Kit
BioControl®, for use with MVP ICON® instruments
About This Item
UNSPSC Code:
41122409
Quality Level
product line
MVP ICON®
manufacturer/tradename
MVP ICON®
application(s)
food and beverages
compatibility
MVP ICON®
General description
MVP ICON® Sample Chamber Field Care Kit (10 Dry Cleaning Swabs)
Application
MVP ICON® field care kit used for cleaning and decontamination of the internal sample chamber area. The MVP ICON® instrument is used for cleaning verification to monitor adherence to both sanitation and HACCP programs. Usage in some environments may introduce liquids, residue, and other contaminants into the sample chamber area, which can potentially affect performance including causing inaccurate ATP readings. This field care kit allows users to clean and decontaminate the internal sample chamber area of the instrument on-site.
Packaging
Clear re-sealable plastic bag
Legal Information
BioControl is a registered trademark of BioControl Systems, Inc.
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?  
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)