Skip to Content
Merck
All Photos(1)

Key Documents

60607

Sigma-Aldrich

Potassium trimethylsilanolate

technical, ≥90% (NT)

Synonym(s):

Trimethylsilanol potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOK
CAS Number:
Molecular Weight:
128.29
Beilstein:
3560282
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

technical

Assay

≥90% (NT)

mp

135-138 °C (lit.)

SMILES string

[K+].C[Si](C)(C)[O-]

InChI

1S/C3H9OSi.K/c1-5(2,3)4;/h1-3H3;/q-1;+1

InChI key

LBKJNHPKYFYCLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diane M Coe et al.
Organic & biomolecular chemistry, 1(7), 1106-1111 (2003-08-21)
A convenient and efficient method for the cleavage of 1,3-oxazolidin-5-ones and 1,3-oxazolidin-2-ones utilising potassium trimethylsilanolate in tetrahydrofuran is described. The benzyloxycarbonyl-protecting group is readily removed under the reaction conditions, whereas the N-benzoyl group is stable. A synthesis of (R)-salmeterol exploiting
Fabricio A Hansel et al.
Rapid communications in mass spectrometry : RCM, 25(13), 1893-1898 (2011-06-04)
A methodology is presented for the determination of dihydroxy fatty acids preserved in the 'bound' phase of organic residues preserved in archaeological potsherds. The method comprises saponification, esterification, silica gel column chromatographic fractionation, and analysis by gas chromatography/mass spectrometry. The
E N M Ho et al.
Analytica chimica acta, 586(1-2), 208-216 (2007-03-28)
Turinabol (4-chloro-17alpha-methyl-17beta-hydroxy-1,4-androstadien-3-one) is a synthetic oral anabolic androgenic steroid. As in the case of other anabolic steroids, it is a prohibited substance in equine sports. The metabolism of turinabol in human has been reported previously; however, little is known about
Grzegorz Piechota et al.
Bioresource technology, 103(1), 16-20 (2011-10-29)
The removal and determination of trimethylsilanol (TMSOH) in landfill gas has been studied before and after the special E3000-ITC System. The system works according to principle of temperature swing. The performance of TMSOH and humidity removal was 20% and more
D M Hui et al.
Clinica chimica acta; international journal of clinical chemistry, 302(1-2), 171-188 (2000-11-14)
We developed a new assay method for fluoride anion (F(-)) a specific metabolite of sarin. Trimethyifluorosilane (TMFS) was derivatized from F(-) with trimethylsilanol, and TMFS was detected with a GC-flame ionization detector (FID) and capillary column system. The linear range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service