Skip to Content
Merck
All Photos(1)

Key Documents

46527

Supelco

19-Norethindrone acetate

VETRANAL®, analytical standard

Synonym(s):

17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one 17-acetate, 17α-Ethynyl-19-nortestosterone 17β-acetate, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one acetate, 19-Norethisterone 17-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C

InChI

1S/C22H28O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,13,17-20H,5-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1

InChI key

IMONTRJLAWHYGT-ZCPXKWAGSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lundström et al.
Maturitas, 70(4), 361-364 (2011-10-01)
To determine the effects of continuous combined hormone therapy, tibolone, black cohosh, and placebo on digitized mammographic breast density in postmenopausal women. A prospective, double-blind, placebo-controlled study of 154 postmenopausal women randomized to estradiol 2 mg/norethisterone acetate 1 mg (E2/NETA)
Donita Africander et al.
Contraception, 84(4), 423-435 (2011-09-17)
Medroxyprogesterone acetate (MPA) and norethisterone (NET) and its derivatives are widely used in female reproductive therapy, but little is known about their mechanisms of action via steroid receptors in the female genital tract. MPA used as a contraceptive has been
Eija Pöllänen et al.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 20(5), 372-379 (2010-08-21)
To investigate the effects of combined hormone replacement therapy (HRT) and its effective agents on the IGF-1 signaling pathway. To examine the effects of HRT on skeletal muscle in vivo, we utilized pre- and post-intervention samples from a randomized double
Simone Ferrero et al.
Reproductive biology and endocrinology : RB&E, 9, 88-88 (2011-06-23)
When aromatase inhibitors are used to treat premenopausal women with endometriosis, additional drugs should be used to effectively down-regulate gonadal estrogen biosynthesis. This randomized prospective open-label study compared the efficacy in treating pain symptoms and the tolerability of letrozole combined
D J Kaser et al.
Journal of pediatric and adolescent gynecology, 25(2), 105-108 (2011-12-14)
To evaluate the efficacy and tolerability of norethindrone acetate (NA) as single-agent hormonal therapy for suppression of endometriosis symptoms in adolescents and young adults. Retrospective study. Two academic medical centers. A keyword search using the query 'NA' was applied to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service