Skip to Content
Merck
All Photos(1)

Documents

46360

Sigma-Aldrich

Flavin adenine dinucleotide disodium salt hydrate

≥90% (HPLC)

Synonym(s):

FAD, FAD-Na2, Riboflavin 5′-adenosine diphosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N9Na2O15P2 · xH2O
CAS Number:
Molecular Weight:
829.51 (anhydrous basis)
Beilstein:
5326842
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥90% (HPLC)

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H]O[H].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6c(N)ncnc56)c2cc1C

InChI

1S/C27H33N9O15P2.2Na.H2O/c1-10-3-12-13(4-11(10)2)35(24-18(32-12)25(42)34-27(43)33-24)5-14(37)19(39)15(38)6-48-52(44,45)51-53(46,47)49-7-16-20(40)21(41)26(50-16)36-9-31-17-22(28)29-8-30-23(17)36;;;/h3-4,8-9,14-16,19-21,26,37-41H,5-7H2,1-2H3,(H,44,45)(H,46,47)(H2,28,29,30)(H,34,42,43);;;1H2/q;2*+1;/p-2/t14-,15+,16+,19-,20+,21+,26+;;;/m0.../s1

InChI key

GXTPHHZYFMAGLX-UJXBNFGUSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

Other Notes

Review

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavin coenzymes: At the crossroads of biological redox chemistry.
C. Walsh
Accounts of Chemical Research, 13, 148-148 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service