Skip to Content
Merck
All Photos(1)

Key Documents

429651

Sigma-Aldrich

Crotonaldehyde-2,4-dinitrophenylhydrazone

analytical standard, for environmental analysis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
250.21
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

grade

analytical standard, for environmental analysis

Quality Level

Assay

99%

form

powder or crystals

mp

192-195 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C\C=C\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C10H10N4O4/c1-2-3-6-11-12-9-5-4-8(13(15)16)7-10(9)14(17)18/h2-7,12H,1H3/b3-2+,11-6+

InChI key

GFUVNGJSSAEZHW-PDGXAJGZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service