Skip to Content
Merck
All Photos(1)

Key Documents

37892

Supelco

Buminafos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H38NO3P
CAS Number:
Molecular Weight:
347.47
Beilstein:
2289568
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

application(s)

agriculture
environmental

storage temp.

2-8°C

SMILES string

CCCCNC1(CCCCC1)P(=O)(OCCCC)OCCCC

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service