Skip to Content
Merck
All Photos(2)

Key Documents

34011

Supelco

2-Nitrobenzaldehyde semicarbazone-13C, 15N2

VETRANAL®, analytical standard

Synonym(s):

2-NP-SCA-13C, 15N2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C713CH8N215N2O3
Molecular Weight:
211.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

mass shift

M+3

SMILES string

N[13C](=O)[15NH]\[15N]=C\c1ccccc1[N+]([O-])=O

InChI

1S/C8H8N4O3/c9-8(13)11-10-5-6-3-1-2-4-7(6)12(14)15/h1-5H,(H3,9,11,13)/b10-5+/i8+1,10+1,11+1

InChI key

OEOKLBSECDAYSM-BNWBFHBJSA-N

Application

2-Nitrobenzaldehyde semicarbazone-13C, 15N2 may be used as an internal standard for the quantification of 4-(2-nitrobenzylidene)semicarbazide (2-NP-SCA) in shrimp samples using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of nitrofuran metabolites and chloramphenicol in shrimp with a single extraction and LC-MS/MS analysis.
El-Demerdash A, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(3), 595-601 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service