Skip to Content
Merck
All Photos(1)

Key Documents

32889

Supelco

Nonylphenol solution

5 μg/mL in acetone, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

5 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

−20°C

InChI

1S/C15H24O/c1-13(2)7-5-3-4-6-8-14-9-11-15(16)12-10-14/h9-13,16H,3-8H2,1-2H3

InChI key

JSFITYFUKSFPBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonylphenol (5 μg/mL in acetone) is grouped under the class of alkylphenol compounds formed from the biodegradation of non-ionic surfactants alkylphenol ethoxylates (APEOs) mainly employed as detergents, emulsifiers and lubricants.

Application

Nonylphenol can be utilized as a reference standard for the quantification of the analyte in water samples by simultaneous derivatization and dispersive liquid-liquid microextraction (DLLME) followed by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shusheng Luo et al.
Journal of chromatography. A, 1217(43), 6762-6768 (2010-07-10)
A simple and fast sample preparation method for the determination of nonylphenol (NP) and octylphenol (OP) in aqueous samples by simultaneous derivatization and dispersive liquid-liquid microextraction (DLLME) was investigated using gas chromatography-mass spectrometry (GC/MS). In this method, a combined dispersant/derivatization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service