Skip to Content
Merck
All Photos(1)

Key Documents

293954

Sigma-Aldrich

Eosin Y

high purity, Dye content 99 %

Synonym(s):

2′,4′,5′,7′-Tetrabromofluorescein, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, Eosin yellowish, Solvent red 43

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H8Br4O5
CAS Number:
Molecular Weight:
647.89
Colour Index Number:
45380:2
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 99%

solubility

spirit: soluble

λmax

524 nm

SMILES string

Oc1c(Br)cc2c(Oc3c(Br)c(O)c(Br)cc3C24OC(=O)c5ccccc45)c1Br

InChI

1S/C20H8Br4O5/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20/h1-6,25-26H

InChI key

DBZJJPROPLPMSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T S Anirudhan et al.
Journal of colloid and interface science, 456, 22-31 (2015-06-19)
A novel adsorbent, 2-mercaptobenzamide modified itaconic acid-grafted-magnetite nanocellulose composite [P(MB-IA)-g-MNCC] was synthesized for adsorbing mercury(II) [Hg(II)] ions selectively from aqueous solutions. Fourier transforms infrared spectroscopy, X-ray diffraction, scanning electron microscopy and thermogravimetric studies were performed to characterize the adsorbent. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service