Skip to Content
Merck
All Photos(1)

Documents

25480

Sigma-Aldrich

1-Chloro-3-nitrobenzene

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
1907947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

bp

236 °C (lit.)

mp

43-47 °C (lit.)
43-47 °C

density

1.534 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(Cl)c1

InChI

1S/C6H4ClNO2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H

InChI key

KMAQZIILEGKYQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hygienic study of the stability of methylnitrophos and pentachloronitrobenzene used in agriculture].
Sh T Atabaev et al.
Gigiena i sanitariia, (2)(2), 74-75 (1983-02-01)
Violeta Simic-Milosevic et al.
Angewandte Chemie (International ed. in English), 48(22), 4061-4064 (2009-04-01)
Chirality chameleon: Inelastic electron tunneling manipulation can be used to change a single chloronitrobenzene (ClNB) molecule, randomly adsorbed on Au(111), into its desired enantiomeric form (r or l, see STM images and ball-and-stick representation) and to vary its rotational orientation.
2-Chloronitrobenzene, 3-chloronitrobenzene and 4-chloronitrobenzene.
IARC monographs on the evaluation of carcinogenic risks to humans, 65, 263-296 (1996-01-01)
V Simic-Milosevic et al.
Physical review letters, 98(11), 116102-116102 (2007-05-16)
A scanning tunneling microscope operating at 5 K is used to induce the isomerization of single chloronitrobenzene molecules on Cu(111) and verify the reaction. The threshold voltage of (227+/-7) mV for this reaction is explained based on electron-induced vibrational heating.
Maofa Ge et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(4), 1499-1502 (2008-06-25)
The electronic structures and substituent effects of o-, m-, and p-chloronitrobenzene and bromonitrobenzene have been studied by ultraviolet photoelectron spectroscopy (UPS). It was found that the o-isomer possesses particular electronic properties. This characteristic depends on the conjugation between the benzene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service