Skip to Content
Merck
All Photos(2)

Documents

18224

Supelco

Fraxetin

analytical standard

Synonym(s):

7,8-Dihydroxy-6-methoxycoumarin, Fraxetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O5
CAS Number:
Molecular Weight:
208.17
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤2.5% water (calc. from elemental analysis)

mp

230-231 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1cc2C=CC(=O)Oc2c(O)c1O

InChI

1S/C10H8O5/c1-14-6-4-5-2-3-7(11)15-10(5)9(13)8(6)12/h2-4,12-13H,1H3

InChI key

HAVWRBANWNTOJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Miguel Nemesio-Gorriz et al.
Journal of experimental botany, 71(19), 6074-6083 (2020-07-01)
Ash dieback, a forest epidemic caused by the invasive fungus Hymenoscyphus fraxineus, threatens ash trees throughout Europe. Within Fraxinus excelsior populations, a small proportion of genotypes show a low susceptibility to the pathogen. We compared the metabolomes from a cohort
Jisoo Song et al.
Pharmaceutics, 13(1) (2021-01-23)
Fraxetin is a coumarin scaffold compound extracted from Fraxinus rhynchophylla. It has antioxidant, anti-inflammatory, hepatoprotective, and antifibrotic effects. Furthermore, fraxetin has anticancer effects in breast and lung cancer. We aimed to evaluate whether fraxetin has anticancer activity in hepatocellular carcinoma
Christopher J Harbort et al.
Cell host & microbe, 28(6), 825-837 (2020-10-08)
Plants benefit from associations with a diverse community of root-colonizing microbes. Deciphering the mechanisms underpinning these beneficial services are of interest for improving plant productivity. We report a plant-beneficial interaction between Arabidopsis thaliana and the root microbiota under iron deprivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service