Skip to Content
Merck
All Photos(1)

Documents

17185

Millipore

m-Enterococcus Agar, modified

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Membrane filter Enterococcus Agar, modified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
cycloheximide, 0.05 g/L
esculin, 1 g/L
nalidixic acid, 0.25 g/L
pancreatic digest of gelatin, 10 g/L
sodium azide, 0.15 g/L
sodium chloride, 15 g/L
yeast extract, 30 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.1±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
water monitoring

microbiology

suitability

slightly selective and differential for Enterococcus spp.

Related Categories

Application

For the recovery of Enterococci in water samples using membrane filter technique along with Esculin Iron Agar for the identification.

Preparation Note

Suspend 71.4 g in 1 litre distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 45°C and aseptically add 15 ml of sterile 1% TTC Solution Cat. No. No. 17779). Mix well and pour into sterile petri plates. Warning: Cycloheximide is very toxic. Avoid skin contact or aerosol formation and inhalation.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service