Skip to Content
Merck
All Photos(1)

Documents

15594

Sigma-Aldrich

Borane tetrahydrofuran complex solution

purum, ~1 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BH3OC4H8
CAS Number:
Molecular Weight:
85.94
Beilstein:
3668402
MDL number:
UNSPSC Code:
12352000

grade

purum

reaction suitability

reagent type: reductant

concentration

~1 M in THF

impurities

≤20% diethylether

density

0.87 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

B.C1CCOC1

InChI

1S/C4H8O.BH3/c1-2-4-5-3-1;/h1-4H2;1H3

InChI key

RMCYTHFAWCWRFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reducing agent; Selective reduction of certain oximes to hydroxylamines; Preparation of chiral boranes as Lewis acid mediators

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.A. Molander et al.
Journal of the American Chemical Society, 114, 2759-2759 (1992)
E.R. Parmee et al.
Journal of the American Chemical Society, 113, 9365-9365 (1991)
F.A.J. Kerdesky et al.
Synthetic Communications, 21, 2203-2203 (1991)
H.C. Brown et al.
Tetrahedron, 35, 567-567 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service