Skip to Content
Merck
All Photos(1)

Key Documents

06429

Supelco

Amberlite IRC50 hydrogen form

weakly acidic, H+-form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

parameter

100 °C max. temp.

moisture

~48%

loss

~45% loss on drying, 110 °C

matrix

acrylic polymer (macroreticular)

particle size

16-50 mesh

operating pH

5-14

capacity

10.0 meq/g by dry weight
3.5 meq/mL by wetted bed volume

Looking for similar products? Visit Product Comparison Guide

Application

Weakly acidic cation exchange resin that selectively adsorbs organic bases such as antibiotics, alkaloids, peptides, and amino acids. Used for purification of many proteins of higher molecular weights. Applied to neutralize reaction mixtures. Good all around use.

Other Notes

For smaller particle size and better efficiency, see Amberlite CG-50 (Type I).

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service