Skip to Content
Merck
All Photos(1)

Key Documents

03910590

(±)-Catechin hydrate

primary reference standard

Synonym(s):

trans-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, trans-3,3′,4′,5,7-Pentahydroxyflavane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O6 · xH2O
CAS Number:
Molecular Weight:
290.27 (anhydrous basis)
Beilstein:
92762
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

Assay

>90.0%

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

storage temp.

−20°C

SMILES string

[H]O[H].O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6.H2O/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7;/h1-5,13,15-20H,6H2;1H2/t13-,15+;/m0./s1

InChI key

OFUMQWOJBVNKLR-NQQJLSKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Catechin hydrate, a flavononol, belongs to the class of flavonoids. It is present in abundance in Siberian larch (Larix sibirica) and in silymarin extract of milk thistle seeds. Its potent chemopreventive action by virtue of the regulation of genes through the ARE-dependent pathway has been widely investigated. It also shows potential antioxidant activity against ovarian cancer cell growth, cellular melanogenesis, human breast cancer, etc.
Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Catechin hydrate may be used as a reference standard for the determination of the analyte in walnut leaves by high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

Polyphenolic antioxidant. Used in traditional Chinese medicine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC determination of phenolic acids, flavonoids and juglone in walnut leaves
Nour V, et al.
Journal of Chromatographic Science, 51(9) (2013)
Stéphane Bastianetto et al.
CNS neuroscience & therapeutics, 15(1), 76-83 (2009-02-21)
Various studies have reported on the neuroprotective effects of polyphenols, widely present in food, beverages, and natural products. For example, we have shown that resveratrol, a polyphenol enriched in red wine and other foods such as peanuts, protects hippocampal cells
Electrochemical, thermodynamic and adsorption studies of (+)-catechin hydrate as natural mild steel corrosion inhibitor in 1 M HCl
Hussin MH and Kassim MJ
International Journal of Electrochemical Science, 6(5), 1396-1414 (2011)
R Hursel et al.
Obesity reviews : an official journal of the International Association for the Study of Obesity, 12(7), e573-e581 (2011-03-04)
Different outcomes of the effect of catechin-caffeine mixtures and caffeine-only supplementation on energy expenditure and fat oxidation have been reported in short-term studies. Therefore, a meta-analysis was conducted to elucidate whether catechin-caffeine mixtures and caffeine-only supplementation indeed increase thermogenesis and
Elena Roura et al.
Clinical chemistry, 52(4), 749-752 (2006-02-14)
Plant polyphenols have been studied largely because of the possibility that they might underlie the protective effects afforded by fruit and vegetable intake against cancer and other chronic diseases. Measurement of polyphenol content excreted in urine as an indicator of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service