Skip to Content
Merck
All Photos(1)

Key Documents

02941

Sigma-Aldrich

Ethylamine

anhydrous, ≥99.0%

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein:
505933
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
Pricing and availability is not currently available.

vapor density

1.56 (15 °C, vs air)

vapor pressure

874 mmHg ( 20 °C)

Assay

≥99.0%

autoignition temp.

721 °F

quality

anhydrous

expl. lim.

14 %
3.5-14 %

impurities

≤0.5% (CH3)2NH
≤0.5% NH3

bp

16.6 °C (lit.)

mp

-81 °C (lit.)

density

0.680 g/mL at 20 °C (lit.)
0.689 g/mL at 25 °C (lit.)

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 250 mL (net ~170 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuying Shuai et al.
Journal of the science of food and agriculture, 90(15), 2563-2567 (2010-08-26)
Theanine, a unique amino acid found almost exclusively in tea plants, has various favourable physiological and pharmacological functions in humans. Gamma-glutamyltranspeptidase (GGT, EC 2.3.2.2) is considered to be the most effective enzyme for the production of theanine. In fact, GGT
Zhixiong Zhong et al.
Analytica chimica acta, 715, 49-56 (2012-01-17)
A new multi-channel purge and trap system coupled with ion chromatography for the determination of six alkylamines in cosmetics was developed. The proposed method, based on purge and trap of the volatile alkylamines, involved in a miniaturization and multi-channel integration
Lisa Vasicek et al.
Analytical chemistry, 81(19), 7876-7884 (2009-09-03)
Electron transfer dissociation (ETD) has proven to be a promising new ion activation method for proteomics applications due to its ability to generate c- and z-type fragment ions in comparison to the y- and b-type ions produced upon the more
Câline Christine et al.
Organic & biomolecular chemistry, 10(46), 9183-9190 (2012-10-23)
The synthesis of a phosphonated acyclic bifunctional chelate L* for the labeling of biomaterial is described. L* is based on a pyridine backbone, functionalized in ortho positions by aminomethyl-bis-methylphosphonic acids, and, in the para position, by a side chain containing
Song Yang et al.
Journal of chromatography. A, 1216(15), 3280-3289 (2009-03-10)
Complementary methods using liquid chromatography-tandem quadrupole mass spectrometry (LC-MS/MS) and comprehensive two-dimensional gas chromatography-time-of-flight mass spectrometry (GCxGC-TOF-MS) were developed and applied to determine targeted metabolites involved in central carbon metabolism [including tricarboxylic acid cycle, serine cycle, ethylmalonyl-coenzyme A (ethylmalonyl-CoA) pathway

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service