Skip to Content
Merck
All Photos(1)

Documents

01390590

Ginkgolic acid C17:1

primary reference standard

Synonym(s):

6-[(10Z)-Heptadecenyl]salicylic acid, Ginkgolic acid II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O3
CAS Number:
Molecular Weight:
374.56
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

SMILES string

CCCCCC\C=C/CCCCCCCCCc1cccc(O)c1C(O)=O

InChI

1S/C24H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18-21-19-17-20-22(25)23(21)24(26)27/h7-8,17,19-20,25H,2-6,9-16,18H2,1H3,(H,26,27)/b8-7-

InChI key

MBYNDKVOZOAOIS-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Other Notes

This compound is commonly found in plants of the genus: ginkgo

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A van Beek et al.
Journal of chromatography. A, 930(1-2), 109-117 (2001-10-30)
A chromatographic procedure for the preparative isolation of six different 6-alkylsalicylic acids (syn. ginkgolic acids) with as alkyl substituents C13:0, C15:0, C15:1, C17:1, C17:2 and, tentatively C17:3 from Ginkgo biloba leaves was developed. The procedure consisted of a combination of
K Ndjoko et al.
Journal of chromatography. B, Biomedical sciences and applications, 744(2), 249-255 (2000-09-19)
Ginkgolic acids (GAs) are toxic phenolic compounds present in the fruits and leaves of Ginkgo biloba L. (Ginkgoacae). Their maximum level in phytopharmaceuticals containing ginkgo extracts has been recently restricted to 5 microg/g by the Commission E of the former

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service