Skip to Content
Merck
All Photos(1)

Key Documents

28-3530

Sigma-Aldrich

Sodium orthosilicate hydrate

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2Na2O · SiO2
Molecular Weight:
184.04 (anhydrous basis)
UNSPSC Code:
12352300
grade:
CP

grade

CP

availability

available only in Japan


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service