Skip to Content
Merck
All Photos(1)

Key Documents

28-1080

Sigma-Aldrich

Silicon standard solution

1000 ppm Si, suitable for atomic absorption spectrometry

Synonym(s):

Silicon dioxide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

concentration

1000 ppm Si

suitability

suitable for atomic absorption spectrometry

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service