11699121001
Roche
PCR Buffer Set
suitable for PCR, pkg of 2 × 2 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 2 × 2 mL
manufacturer/tradename
Roche
technique(s)
PCR: suitable
storage temp.
−20°C
General description
The kit-provided PCR buffer without MgCl2, in combination with the kit-provided MgCl2 stock solution, are used for the individual adjustment of the Mg2+ concentration. In most PCR applications, a concentration of 1.5 mM MgCl2 will yield satisfactory results with a dNTP concentration of 200 μM for each nucleotide. To optimize the PCR, titrate to determine the optimal Mg2+ concentration for high PCR specificity and amplicon yield.
Application
For optimization of the magnesium-chloride concentration in the polymerase chain reaction (PCR) using Taq DNA Polymerase.
Packaging
1 set containing 2 components
1 set containing 3 components
Quality
Function test: Each lot of buffer is tested in PCR.
Absence of contaminants: No exo- or endonuclease activity is detectable according to the current Quality Control procedures.
Absence of contaminants: No exo- or endonuclease activity is detectable according to the current Quality Control procedures.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Kit Components Only
Product No.
Description
- MgCl2 Stock Solution 25 mM
- PCR Buffer without Mg2+ 10x concentrated
- Alkaline Phosphatase Substrate, CSPD
- Enhancer, Emerald II
- Assay Buffer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service