Skip to Content
Merck
All Photos(1)

Documents

10108677001

Roche

Poly(A) x (dT)15

Polyadenylic acid x pentadecathymidylic acid

Synonym(s):

PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

Quality Level

form

lyophilized

mol wt

660 kDa

packaging

pkg of 5 U

manufacturer/tradename

Roche

absorbance ratio

A290/260 nm 0.13-0.17
A280/260 nm 0.48-0.52
A250/260 nm 0.78-0.82

storage temp.

−20°C (−15°C to −25°C)

Related Categories

General description

Poly (A) x (dT)15 is used as a template of reverse transcriptases and, for example, can be applied to test the activity of reverse transriptases.
Contents: Lyophilized with NaCl, base ratio 1:1.15.

Application

Poly(A) x (dT)15 is used as a primer for Reverse Transcriptase.

Quality

Absorbance determination A250/A260, A280/A260, A290/A260

Sequence

Chain Length approx. 1,000 bp

Unit Definition

1 A260 unit equals 50 μg

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service