Key Documents
ZFCBEQXAU
Milli-Q
Country box Milli-Q® EQ 7008/16 AU
Available only in Austria
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
compatibility
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
General description
The Country box for the Milli-Q® EQ 7008/16 system is an installation accessory that contains a printed quick reference guide, a user manual in local language and in English on a USB key, and a power cord adapted to the local electrical network.
Application
The components of the Country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany