Skip to Content
Merck
All Photos(1)

Key Documents

S26F-MLSG

Sigma-Aldrich

Goat Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.71

antibody form

serum

Mode of action

| inhibits

General description

Normal goat serum from healthy animals collected from USDA inspected facilities. All animals have received ante and post mortem inspections and were found to be free of contagious diseases. <br /><br />HEMOGLOBIN: ~3.5 mg/dL <br /><br />COUNTRY OF ORIGIN: USA

Application

Blocking reagent. Dilute as needed with PBS or other buffer. Typical working concentrations are 0.1-5% serum in buffer. For best results, it is recommended to filter the diluted serum solution through a 0.2-0.45μm membrane prior to use.Blocking reagent; dilute as needed with PBS or other buffer. Typical working concentrations are 0.1-5% serum in buffer.

Optimal dilutions determined by enduser.

Physical form

Liquid clean serum, unfiltered, containing no preservatives.

Storage and Stability

Store frozen at -20°C in undiluted aliquots for up to twelve months. Avoid repeated freeze/thaw cycles.

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: [email protected]
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service