Skip to Content
Merck
All Photos(1)

Key Documents

PC420

Sigma-Aldrich

Anti-Capsaicin Receptor (Ab-1) (824-838) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Vanilloid Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Trpv1(193034)

General description

Anti-Capsaicin Receptor (Ab-1) (824-838), rabbit polyclonal, recognizes the ~100 kDa capsaicin receptor in mouse spinal cord extract. It is validated for WB, IF & IHC in frozen and paraffin sections.
Protein A purified rabbit polyclonal antibody. Recognizes the ~100 kDa capsaicin receptor protein.
Recognizes the ~100 kDa capsaicin receptor protein in mouse spinal cord extract.

Immunogen

a synthetic peptide (EDAEVFKDSMVPGEK) corresponding to amino acids 824-838 of rat capsaicin receptor, conjugated to KLH

Application



Frozen Sections (5 g/ml, see application references)
Immunoblotting (5-10 g/ml)
Immunofluorescence (2 g/ml)
Paraffin Sections (5 g/ml; no pre-treatment required)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Positive Control
Mouse spinal cord dorsal horn

Other Notes

Caterina, M.J., et al. 1999. Nature398, 436.
Gau, A., et al. 1999. Eur. J. Neurosci.11, 946.
Michael, G.J., and Priestley, J.V. 1999. J. Neurosci.19, 1844.
Caterina, M.J., et al. 1997. Nature389, 816.
Szallasi, A. 1994. Gen. Pharmacol.25, 223.
Beven, S., and Szolcsanyi, J. 1990. Trends Pharmacol. Sci.11, 330.
Fields, H.L. 1987. Pain (New York: McGraw-Hill).
Immunohistochemistry was performed on mouse spinal cord sections fixed in 4% paraformaldehyde. Immunohistochemical staining results correlate well with other independent reports and in situ hybridization studies. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N J Spencer et al.
Neuroscience, 153(2), 518-534 (2008-04-09)
Rodents detect visceral pain in response to noxious levels of rectal distension. However, the mechanoreceptors that innervate the rectum and respond to noxious levels of rectal distension have not been identified. Here, we have identified the mechanoreceptors of capsaicin-sensitive rectal
Klaus Bielefeldt et al.
American journal of physiology. Gastrointestinal and liver physiology, 291(5), G987-G997 (2006-05-27)
Recent studies suggest that the capsaicin receptor [transient receptor potential vanilloid (TRPV)1] may play a role in visceral mechanosensation. To address the potential role of TRPV1 in vagal sensory neurons, we developed a new in vitro technique allowing us to
M Kawashima et al.
European journal of histochemistry : EJH, 56(2), e21-e21 (2012-06-13)
Transient receptor potential vanilloid subfamily member 1 (TRPV1) is activated by capsaicin, acid, and heat and mediates pain through peripheral nerves. In the tongue, TRPV1 expression has been reported also in the epithelium. This indicates a possibility that sensation is
Nathaniel A Jeske et al.
Pain, 146(3), 301-307 (2009-09-22)
Post-translational modifications on various receptor proteins have significant effects on receptor activation. For the Transient Receptor Potential family V type 1 (TRPV1) receptor, phosphorylation of certain serine/threonine amino acid residues sensitizes the receptor to activation by capsaicin and heat. Although
Chiao-Chi Chen et al.
Scientific reports, 9(1), 12348-12348 (2019-08-28)
Gout is one of the most painful disease conditions. The central mechanism of pain processing in this condition remains elusive. Cerebral blood volume (CBV) responses are faithful correlates of brain activity changes; the application of CBV-weighted functional magnetic resonance imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service