Skip to Content
Merck
All Photos(2)

Documents

MABT345

Sigma-Aldrich

Anti-Prelamin-A Antibody, clone 7G11

clone 7G11, from rat

Synonym(s):

Prelamin-A/C, Prelamin-A, Lamin-A/C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7G11, monoclonal

species reactivity

mouse

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lmna(16905)

General description

Lamin-A is a key component of the nuclear lamina, a protein network underlying the inner membrane that determines nuclear shape and size. The same gene (Lmna, Dhe, or Lamn1 in murine species; Gene ID 16905) that encodes lamin-A also codes for the alternatively spliced prelamin-C isoforms (UniProt P48678-2 and P48678-3). Murine lamin-A (UniProt P48678-1) is initially produced as a 665-amino acid prelamin A that terminals in a CAAX motif. It is further processed post-translationally by four enzymatic steps to yield the mature Lamin-A. Farnesylation of Cys662, endoproteolytic cleavage of the last three amino acids (a.a. 663-665), carboxylmethylation of Cys662, and the final endoproteolytic removal of the remaining propeptide sequence (a.a. 648-662), including the farnesylated Cys662, is catalyzed by the ER membrane zinc metalloproteinase ZMPSTE24.

Specificity

Clone 7G11 detects prelamin-A regardless of its farnesylation state, but not mature Lamin-A (a.a. 1-647) lacking the C-terminal propeptide sequence. Clone 7G11 does not detect the pre- or mature forms of the spliced isoforms C1 & C2 (Lamin-C; P48678-2 & P48678-3).

Immunogen

Epitope: C-terminal propeptide sequence
Linear peptide corresponding to the C-terminal propeptide sequence of mouse Prelamin-A.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Prelamin-A Antibody, clone 7G11 is validated for use in Western Blotting and Immunofluorescence for the detection of Prelamin-A.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected 2-day FTI (5 µM; Cat. No. 344550) treatment-induced prelamin-A accumulation in C2C12 cell lysate.
Immunofluorescence Analysis: A representative lot detected upregulated prelamin-A immunoreactivity in keratinocytes, but not in dermal fibroblasts by fluorescent immunohistochemistry using paraformaldehyde-fixed and paraffin-embedded skin sections from keratinocyte-specific Fntb knockout mice, while prelamin-A upregulation was seen in both keratinocytes and dermal fibroblasts from mice with non-tissue/cell type-specific Zmpste24 knockout (Lee, R., et al. (2010). Hum. Mol. Genet.19(8):1603-1617).
Immunofluorescence Analysis: A representative lot stained the nuclear rim of hepatocytes by fluorescent immunohistochemistry using methanol-fixed and Tween-permeabilized frozen liver sections from a transgenic mouse strain that produces the C662S non-farnesylated prelamin-A only (nPLAO/nPLAO) and no lamin-C, while cellular prelamin-A level was undetectable in liver sections from wild-type mice or a mouse strain that produces wild-type prelamin-A only (PLAO/PLAO) and no lamin-C (Davies, B.S., et al. (2010). Hum. Mol. Genet. 19(13):2682-2694).
Western Blotting Analysis: A representative lot detected cellular prelamin-A accumulation upon farnesyltransferase inhibitor (FTI) treatment in murine fibroblasts. Clone 7G11 selectively detects prelamin-A, but not mature lamin-A or lamin-C (Lee, R., et al. (2010). Hum. Mol. Genet.19(8):1603-1617).
Note: Under normal condition, cellular prelamin-A level is below detection. Any newly produced prelamin-A is quickly processed further to yield mature lamin-A. Cellular farnesyltransferase and/or ZMPSTE24 activity must be inhibited (by gene knockout or inhibitor treatment) to allow antibody-based detection of cellular prelamin-A.

Quality

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected 2-day FTI-276 (5 µM; Cat. No. 344550) treatment-induced prelamin-A accumulation in murine RAW 264.7 macrophages.

Target description

~74 kDa observed. This antibody detects A form prelamin but does not detect isoform C.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Casasola et al.
Nucleus (Austin, Tex.), 7(1), 84-102 (2016-02-24)
Lamin A is part of a complex structural meshwork located beneath the nuclear envelope and is involved in both structural support and the regulation of gene expression. Lamin A is initially expressed as prelamin A, which contains an extended carboxyl
Carlos González-Blanco et al.
Aging, 14(5), 2047-2061 (2022-03-21)
Hutchinson-Gilford Progeria Syndrome is an ultrarare disease which is characterized by an accelerated senescence phenotype with deleterious consequences to people suffering this pathology. The production of an abnormal protein derived from lamin A, called progerin, presents a farnesylated domain, which
Xue Chen et al.
eLife, 10 (2021-02-03)
A farnesylated and methylated form of prelamin A called progerin causes Hutchinson-Gilford progeria syndrome (HGPS). Inhibiting progerin methylation by inactivating the isoprenylcysteine carboxylmethyltransferase (ICMT) gene stimulates proliferation of HGPS cells and improves survival of Zmpste24-deficient mice. However, we don't know

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service