Skip to Content
Merck
All Photos(2)

Key Documents

MABS1287

Sigma-Aldrich

Anti-EPO-R Antibody, clone VP-2E8

clone VP-2E8, from mouse

Synonym(s):

Erythropoietin Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VP-2E8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... EPOR(2057)

General description

Erythropoietin receptor (UniProt: P19235, also known as EPO-R) is encoded by the EPOR gene (Gene ID: 2057) in human. EPO-R is a single-pass type I membrane protein that serves as a receptor for erythropoietin. It mediates erythropoietin-induced erythroblast proliferation and differentiation. Upon EPO stimulation, EPO-R dimerizes triggering the JAK2/STAT5 signaling cascade. EPO-R has an extracellular domain (aa 25-250), a transmembrane domain (aa 251-273), and a cytoplasmic domain (aa 274-508). It is synthesized with a signal peptide (aa 1-24) that is cleaved to generate mature EPO-R. Upon stimulation following Erythropoietin (EPO) binding, EPO-R is phosphorylated on C-terminal tyrosine residues by JAK2. The phosphotyrosine motifs are also recruitment sites for several SH2-containing proteins and adapter proteins, which mediate cell proliferation. Phosphorylation on Tyrosine 454 is shown to be essential for PTPN6 interaction, and on Tyrosine 426 for PTPN11 interaction. Tyrosine 426 is also required for SOCS3 binding, however, Tyr-454/Tyr-456 motif is reported to be the preferred binding site. Ubiquitination of EPO-R at Lysine 81 is required for receptor internalization, whereas ubiquitination at Lysine 453 promotes trafficking of activated receptors to the lysosomes for degradation. Defects in EPOR gene are linked to familial erythrocytosis which is characterized by increased serum red blood cell mass, elevated hemoglobin and hematocrit, and hypersensitivity of erythroid progenitors to erythropoietin.

Specificity

Clone VP-2E8 recognizes EPO-R in human cells. It targets an epitope within the extracellular domain.

Immunogen

A recombinant fragment corresponding to the extracellular domain of human EPO-R.
Epitope: extracellular domain

Application

Anti-EPO-R, clone VP-2E8, Cat. No. MABS1287, is a mouse monoclonal antibody that detects erythropoietin receptor and is tested for use in Flow Cytometry, Immunocytochemistry, Immunofluorescence, and Immunoprecipitation.
Flow Cytometry Analysis: 2 µg/mL from a representative lot detected EPO-R in UT-7 (EPO dependent) cells.

Immunoprecipitation Analysis: A representative lot detected EPO-R in Immunoprecipitation applications (Maxwell, P., et. al. (2015). Br J Haematol. 168(3):429-42).

Immunofluorescence Analysis: A representative lot detected EPO-R in Immunofluorescence applications (Maxwell, P., et. al. (2015). Br J Haematol. 168(3):429-42).

Flow Cytometry Analysis: A representative lot detected EPO-R in Flow Cytometry applications (Maxwell, P., et. al. (2015). Br J Haematol. 168(3):429-42).
Research Category
Signaling

Quality

Evaluated by Immunocytochemistry in starved UT-7 cells.

Immunocytochemistry Analysis: 1:500 dilution of this antibody detected EPO-R in starved UT-7 cells.

Target description

55.06 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service